Order Entry
Spain
ContactUsLinkComponent
Anti-Mouse IgG1 IgG2A IgG2B IgG3 IgA IgM IgD Rabbit Polyclonal Antibody
Anti-Mouse IgG1 IgG2A IgG2B IgG3 IgA IgM IgD Rabbit Polyclonal Antibody
Catalog # BIRBORB22382-1
Supplier:  Biorbyt
Anti-Mouse IgG1 IgG2A IgG2B IgG3 IgA IgM IgD Rabbit Polyclonal Antibody
Catalog # BIRBORB22382-1
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgA/IgD/IgG1/IgG2a/IgG2b/IgG3/IgM
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Format:
    antiserum
  • Form:
    lyophilized
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Purified polyclonal Mouse IgG and pools of homogenous IgG, IgA, IgM and IgD Freunds complete adjuvant is used in the first step of the immunization procedure.
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Type: Secondary
Antigen: Mouse IgG1 IgG2A IgG2B IgG3 IgA IgM IgD
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: