Anti-Malic Dehydrogenase Rabbit Polyclonal Antibody
BIRBORB21015-10
: Biorbyt
- Antibody type:Primary
- Antigen name:Malate Dehydrogenase
- Antigen symbol:MDH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Storage buffer:phosphate buffered saline
- Storage temperature:Cool
- Concentration:1 mg/ml, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Malic dehydrogenase is isolated and purified from human erythrocytes. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:10 mg
- Pk:10 mg
Type: Primary
Antigen: Malic Dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human