Order Entry
Spain
ContactUsLinkComponent
Anti-3-Phosphoglyceric Phosphokinase Rabbit Polyclonal Antibody
Anti-3-Phosphoglyceric Phosphokinase Rabbit Polyclonal Antibody
Catalog # BIRBORB20869-10
Supplier:  Biorbyt
Anti-3-Phosphoglyceric Phosphokinase Rabbit Polyclonal Antibody
Catalog # BIRBORB20869-10
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    3-Phosphoglyceric Phosphokinase
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Yeast
  • Storage buffer:
    phosphate buffered saline
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml, no foreign proteins added,
  • Shipping temperature:
    Ice
  • Immunogen:
    3-Phosphoglyceric phosphokinase isolated and purified from bakers yeast. Freunds complete adjuvant is used in the first step of the immunization procedure.
  • Size:
    10 mg
  • Pk:
    10 mg

Specifications

About this item

Type: Primary
Antigen: 3-Phosphoglyceric Phosphokinase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Yeast