Anti-Nuclease S1 Rabbit Polyclonal Antibody
Catalog # BIRBORB20849-10
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:Nuclease S1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Storage buffer:phosphate buffered saline
- Storage temperature:Cool
- Concentration:1 mg/ml, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Nuclease S1 isolated and purified from Aspergillus oryzae. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:10 mg
- Pk:10 mg
Specifications
About this item
Type: Primary
Antigen: Nuclease S1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: