Order Entry
Spain
ContactUsLinkComponent
 
Anti-IgG2b/IgG2a Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin)
 :  Biorbyt
 
undefined
Anti-IgG2b/IgG2a Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin)
 :  Biorbyt

 

  • Catalog No:
  • BIRBORB12209-50
  • BIRBORB12209-20
  • Antigen symbol:
  • IgG2b/IgG2a
  • IgG2b/IgG2a
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG2b, IgG2a
  • IgG2b, IgG2a
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Purification:
  • purified by affinity chromatography
  • purified by affinity chromatography
  • Storage buffer:
  • Phosphate-buffered solution, pH 7,4, containing 0,09% sodium azide and 0,2% (w/v) BSA
  • Phosphate-buffered solution, pH 7,4, containing 0,09% sodium azide and 0,2% (w/v) BSA
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

Frequently Bought Together