Order Entry
Spain
ContactUsLinkComponent
Anti-CD3/CD16+56 Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin)
Anti-CD3/CD16+56 Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin)
Supplier:  Biorbyt
Certificates
Anti-CD3/CD16+56 Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin)
Supplier:  Biorbyt

Specifications

  • Catalog No:
  • BIRBORB12157-20
  • BIRBORB12157-50
  • Antigen symbol:
  • CD3/CD16/CD56
  • CD3/CD16/CD56
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG2a, IgG1
  • IgG2a, IgG1
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Purification:
  • purified by affinity chromatography
  • purified by affinity chromatography
  • Storage buffer:
  • Phosphate-buffered solution 0,09% sodium azide and 0,2% (w/v) BSA
  • Phosphate-buffered solution 0,09% sodium azide and 0,2% (w/v) BSA
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together