Anti-EMP1 Rabbit polyclonal antibody
Supplier: Biorbyt
Certificates
About this item
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: EMP1 (epithelial membrane protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- BIRBORB10588-100
- BIRBORB10588-200
- Antigen symbol:
- EMP1
- EMP1
- Antigen name:
- Epithelial membrane protein 1
- Epithelial membrane protein 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide
- Synthetic peptide
- Form:
- liquid
- liquid
- Molecular weight:
- 17 kDa
- 17 kDa
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Concentration:
- 0,5 mg/ml
- 0,5 mg/ml
- Storage buffer:
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together