Order Entry
Spain
ContactUsLinkComponent
Anti-IFNK Mouse Monoclonal Antibody [clone: 1B7]
Anti-IFNK Mouse Monoclonal Antibody [clone: 1B7]
Catalog # ABNOH00056832-M01
Proveedor:  Abnova
undefined
Anti-IFNK Mouse Monoclonal Antibody [clone: 1B7]
Catalog # ABNOH00056832-M01
Proveedor:  Abnova

Especificaciones

  • Antibody type:
    Primary
  • Antigen name:
    Interferon kappa
  • Antigen symbol:
    IFNK
  • Clonality:
    Monoclonal
  • Clone:
    1B7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    56832
  • Antigen synonyms:
    IFNT1|INFE1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Especificaciones

Acerca de este artículo

Type: Primary
Antigen: IFNK
Clonality: Monoclonal
Clone: 1B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human