Anti-LRAT Mouse Monoclonal Antibody [clone: 4E12]
Catalog # ABNOH00009227-M05
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Lecithin retinol acyltransferase
- Antigen symbol:LRAT
- Clonality:Monoclonal
- Clone:4E12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9227
- Antigen synonyms:Lecithin retinol acyltransferase|Phosphatidylcholine--retinol O-acyltransferase|LCA14|Phosphatidylcholine-retinol O-acyltransferase
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: LRAT
Clonality: Monoclonal
Clone: 4E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human