Anti-TNNC1 Mouse Monoclonal Antibody [clone: 1F8-A9]
ABNOH00007134-M01
: Abnova
- Antibody type:Primary
- Antigen name:Troponin C type 1
- Antigen symbol:TNNC1
- Clonality:Monoclonal
- Clone:1F8-A9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:7134
- Antigen synonyms:TNNC|TNC|TN-C|CMD1Z|CMH13
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: TNNC1
Clonality: Monoclonal
Clone: 1F8-A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human