Anti-STAC Rabbit Polyclonal Antibody
Catalog # ABNOH00006769-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:SH3 and cysteine rich domain
- Antigen symbol:STAC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6769
- Antigen synonyms:STAC1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: STAC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat