Anti-SPRR3 Mouse Monoclonal Antibody [clone: 4A12]
ABNOH00006707-M01
: Abnova
- Antibody type:Primary
- Antigen name:Small Proline-Rich Protein 3
- Antigen symbol:SPRR3
- Clonality:Monoclonal
- Clone:4A12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6707
- Antigen synonyms:22 kDa pancornulin|Small proline rich protein 3|Small proline-rich protein 3|SPRC|Cornin beta|Cornin B|Sprr3|SPRR3_HUMAN.|Esophagin
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: SPRR3
Clonality: Monoclonal
Clone: 4A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human