Anti-SFTPD Mouse Polyclonal Antibody
Catalog # ABNOH00006441-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Surfactant Protein D
- Antigen symbol:SFTPD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6441
- Antigen synonyms:COLEC7|SFTP4|SP-D|PSP-D
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SFTPD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human