Order Entry
Spain
ContactUsLinkComponent
Anti-PYCR1 Rabbit Polyclonal Antibody
Anti-PYCR1 Rabbit Polyclonal Antibody
Catalog # ABNOH00005831-D01
Supplier:  Abnova
undefined
Anti-PYCR1 Rabbit Polyclonal Antibody
Catalog # ABNOH00005831-D01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Pyrroline-5-carboxylate reductase 1
  • Antigen symbol:
    PYCR1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    5831
  • Antigen synonyms:
    PIG45|ARCL3B|P5CR|PYCR|P5C|PP222|PRO3|ARCL2B
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: PYCR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human