Anti-PPP2R2C Mouse Monoclonal Antibody [clone: 6D1]
ABNOH00005522-M01
: Abnova
- Antibody type:Primary
- Antigen name:Protein Phosphatase 2, Regulatory Subunit 2C
- Antigen symbol:PPP2R2C
- Clonality:Monoclonal
- Clone:6D1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5522
- Antigen synonyms:PR52|IMYPNO|IMYPNO1|B55-GAMMA|PR55G
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: PPP2R2C
Clonality: Monoclonal
Clone: 6D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human