Anti-NACA (nascent Polypeptide-associated Complex alpha Subunit) Mouse Polyclonal Antibody
Catalog # ABNOH00004666-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:NACA (nascent Polypeptide-associated Complex alpha Subunit)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:4666
- Antigen synonyms:skNAC|HSD48|NACA1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: NACA (nascent Polypeptide-associated Complex alpha Subunit)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human