Anti-MARS Rabbit Polyclonal Antibody
ABNOH00004141-D01
: Abnova
- Antibody type:Primary
- Antigen name:Methionyl tRNA synthetase
- Antigen symbol:MARS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Cross adsorption:No
- Gene ID:4141
- Antigen synonyms:MRS|SPG70|MTRNS|CMT2U|METRS|ILLD|ILFS2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: MARS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human