Anti-CD1D Mouse Polyclonal Antibody
Catalog # ABNOH00000912-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CD1d molecule
- Antigen symbol:CD1D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:912
- Antigen synonyms:T-cell surface glycoprotein CD1d1|Ly-38|CD1.1|AI747460|Cd1d|Cd1a|antigen-presenting glycoprotein CD1d1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: CD1D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human