Order Entry
Spain
ContactUsLinkComponent
Anti-FCGR2/3 Rat monoclonal antibody [clone: 2.4G2]
Anti-FCGR2/3 Rat monoclonal antibody [clone: 2.4G2]
Supplier:  Tonbo Biosciences
Certificates
Anti-FCGR2/3 Rat monoclonal antibody [clone: 2.4G2]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 70-0161-U500
  • 70-0161-M001
  • 70-0161-U100
  • Antigen symbol:
  • CD16 + CD32
  • CD16 + CD32
  • CD16 + CD32
  • Antigen name:
  • FcR IgG Low Affinity 2
  • FcR IgG Low Affinity 2
  • FcR IgG Low Affinity 2
  • Antigen synonyms:
  • CD32|Fc-gamma-RIIc|Fc gamma Receptor III|Fc-gamma RII-b|Fc gamma Receptor II|FcRII-c|FCGR3B|Fc-gamma-RIIb|FCGR3A|FCG2|Low affinity immunoglobulin gamma Fc region receptor II-c|CD32 receptor 2|Fc gamma Receptor 2|FCRIII|CD16A|Fc gamma Receptor 3|Fc-gamma RII-c|RP11-5K23.1|FCGR3|CDw32|FCRIIIA|FCR-10|FCGR2B|FCGR2|CD16B|FCGR2A|FCGRIII|FCGR2C|FcRII-b|Low affinity immunoglobulin gamma Fc region receptor II-b
  • CD32|Fc-gamma-RIIc|Fc gamma Receptor III|Fc-gamma RII-b|Fc gamma Receptor II|FcRII-c|FCGR3B|Fc-gamma-RIIb|FCGR3A|FCG2|Low affinity immunoglobulin gamma Fc region receptor II-c|CD32 receptor 2|Fc gamma Receptor 2|FCRIII|CD16A|Fc gamma Receptor 3|Fc-gamma RII-c|RP11-5K23.1|FCGR3|CDw32|FCRIIIA|FCR-10|FCGR2B|FCGR2|CD16B|FCGR2A|FCGRIII|FCGR2C|FcRII-b|Low affinity immunoglobulin gamma Fc region receptor II-b
  • CD32|Fc-gamma-RIIc|Fc gamma Receptor III|Fc-gamma RII-b|Fc gamma Receptor II|FcRII-c|FCGR3B|Fc-gamma-RIIb|FCGR3A|FCG2|Low affinity immunoglobulin gamma Fc region receptor II-c|CD32 receptor 2|Fc gamma Receptor 2|FCRIII|CD16A|Fc gamma Receptor 3|Fc-gamma RII-c|RP11-5K23.1|FCGR3|CDw32|FCRIIIA|FCR-10|FCGR2B|FCGR2|CD16B|FCGR2A|FCGRIII|FCGR2C|FcRII-b|Low affinity immunoglobulin gamma Fc region receptor II-b
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Clone:
  • 2.4G2
  • 2.4G2
  • 2.4G2
  • Reactivity:
  • Mouse
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Rat
  • Gene ID:
  • 14130/14131
  • 14130/14131
  • 14130/14131
  • Isotype:
  • IgG2b
  • IgG2b
  • IgG2b
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together