Order Entry
Spain
ContactUsLinkComponent
Anti-IL2RA Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: PC61.5]
Anti-IL2RA Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: PC61.5]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-IL2RA Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: PC61.5]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 60-0251-U100
  • 60-0251-U025
  • Antigen symbol:
  • IL2RA
  • IL2RA
  • Antigen name:
  • Interleukin 2 Receptor Alpha
  • Interleukin 2 Receptor Alpha
  • Antigen synonyms:
  • TCGFR|IL2-RA|p55|IL-2R subunit alpha|IL-2 receptor subunit alpha|IL-2-RA|IL2R|TAC antigen|IDDM10|CD25
  • TCGFR|IL2-RA|p55|IL-2R subunit alpha|IL-2 receptor subunit alpha|IL-2-RA|IL2R|TAC antigen|IDDM10|CD25
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)/Cy7®
  • PE (Phycoerythrin)/Cy7®
  • Clone:
  • PC61.5
  • PC61.5
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 16184
  • 16184
  • Isotype:
  • IgG1 lambda
  • IgG1 lambda
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together