Order Entry
Spain
ContactUsLinkComponent
Anti-TRB Armenian Hamster Monoclonal Antibody (FITC (Fluorescein)) [clone: H57-597]
Anti-TRB Armenian Hamster Monoclonal Antibody (FITC (Fluorescein)) [clone: H57-597]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-TRB Armenian Hamster Monoclonal Antibody (FITC (Fluorescein)) [clone: H57-597]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 35-5961-U025
  • 35-5961-U100
  • 35-5961-U500
  • Antigen symbol:
  • TCRB
  • TCRB
  • TCRB
  • Antigen name:
  • T-cell Receptor beta
  • T-cell Receptor beta
  • T-cell Receptor beta
  • Antigen synonyms:
  • TCRB|T cell receptor beta|TRB|T cell antigen receptor
  • TCRB|T cell receptor beta|TRB|T cell antigen receptor
  • TCRB|T cell receptor beta|TRB|T cell antigen receptor
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • H57-597
  • H57-597
  • H57-597
  • Reactivity:
  • Mouse
  • Mouse
  • Mouse
  • Host:
  • Armenian hamster
  • Armenian hamster
  • Armenian hamster
  • Gene ID:
  • 21577
  • 21577
  • 21577
  • Isotype:
  • IgG
  • IgG
  • IgG
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together