Order Entry
Spain
ContactUsLinkComponent
 
Anti-TER-119 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: TER-119]
 
undefined
Anti-TER-119 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: TER-119]

 

  • Catalog No:
  • 35-5921-U100
  • 35-5921-U500
  • 35-5921-U025
  • Antigen symbol:
  • LY76
  • LY76
  • LY76
  • Antigen name:
  • Lymphocyte Antigen 76
  • Lymphocyte Antigen 76
  • Lymphocyte Antigen 76
  • Antigen synonyms:
  • Ter119|TER-119
  • Ter119|TER-119
  • Ter119|TER-119
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • TER-119
  • TER-119
  • TER-119
  • Reactivity:
  • Mouse
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Rat
  • Gene ID:
  • 104231
  • 104231
  • 104231
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • IgG2b kappa
  • Western blot:
  • Yes
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice
  • Ice

 

Frequently Bought Together