Order Entry
Spain
ContactUsLinkComponent
 
Anti-CD3E Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 17A2]
 
undefined
Anti-CD3E Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 17A2]

 

  • Catalog No:
  • 35-0032-U100
  • 35-0032-U025
  • Antigen symbol:
  • CD3G
  • CD3G
  • Antigen name:
  • CD3 antigen, gamma polypeptide
  • CD3 antigen, gamma polypeptide
  • Antigen synonyms:
  • T3g|Ctg3|Ctg-3
  • T3g|Ctg3|Ctg-3
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • 17A2
  • 17A2
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 12502
  • 12502
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

Frequently Bought Together