Anti-SPRY4 Rabbit Polyclonal Antibody
Catalog # ABGEAP7446D
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Sprouty RTK Signaling Antagonist 4
- Antigen symbol:SPRY4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:Spry-4|Protein sprouty homolog 4|SPRY4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: SPRY4 (sprouty homolog 4 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human