Anti-KRT15 Rabbit Polyclonal Antibody
Catalog # AVIVAVARP00005P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:keratin 15, type I
- Antigen symbol:KRT15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:3866
- Antigen synonyms:K1CO|CK15|K15
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: KRT15 (keratin 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: