Anti-CTTNBP2NL Rabbit Polyclonal Antibody
AVIVARP68240_P050
- Antibody type:Primary
- Antigen name:CTTNBP2 N-terminal like
- Antigen symbol:CTTNBP2NL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:55917
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: CTTNBP2NL (CTTNBP2 N-terminal like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human