Anti-EEFSEC Rabbit Polyclonal Antibody
Catalog # AVIVARP65659_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:eukaryotic elongation factor, selenocysteine-tRNA-specific
- Antigen symbol:EEFSEC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:60678
- Antigen synonyms:SELB|EFSEC
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: EEFSEC (eukaryotic elongation factor, selenocysteine-tRNA-specific)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human