Anti-CAPRIN1 Rabbit Polyclonal Antibody
AVIVARP65220_P050
- Antibody type:Primary
- Antigen name:Cell cycle associated protein 1
- Antigen symbol:CAPRIN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:4076
- Antigen synonyms:GPIP137|M11S1|RNG105|GRIP137|p137GPI|GPIAP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: CAPRIN1 (cell cycle associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human