Order Entry
Spain
ContactUsLinkComponent
Anti-GPRC6A Rabbit Polyclonal Antibody
Anti-GPRC6A Rabbit Polyclonal Antibody
Catalog # AVIVARP64456_P050
undefined
Anti-GPRC6A Rabbit Polyclonal Antibody
Catalog # AVIVARP64456_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    G Protein-Coupled Receptor, Class C, Group 6, Member A
  • Antigen symbol:
    GPRC6A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Gene ID:
    222545
  • Antigen synonyms:
    GPCR|bA86F4.3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: GPRC6A (G protein-coupled receptor, class C, group 6, member A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human