Anti-MSL1 Rabbit Polyclonal Antibody
Catalog # AVIVARP64394_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:male-specific lethal 1 homolog (Drosophila)
- Antigen symbol:MSL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:339287
- Antigen synonyms:hMSL1|MSL-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: MSL1 (male-specific lethal 1 homolog (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: