Anti-PDE4C Rabbit Polyclonal Antibody
AVIVARP64361_P050
- Antibody type:Primary
- Antigen name:phosphodiesterase 4C, cAMP-specific
- Antigen symbol:PDE4C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:5143
- Antigen synonyms:DPDE1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: PDE4C (phosphodiesterase 4C, cAMP-specific)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: