Anti-PLCXD1 Rabbit Polyclonal Antibody
Catalog # AVIVARP62895_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:phosphatidylinositol-specific phospholipase C, X domain containing 1
- Antigen symbol:PLCXD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:55344
- Antigen synonyms:LL0XNC01-136G2.1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: PLCXD1 (phosphatidylinositol-specific phospholipase C, X domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human