Anti-NSL1 Rabbit Polyclonal Antibody
Catalog # AVIVARP62184_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:NSL1, MIS12 kinetochore complex component
- Antigen symbol:NSL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:25936
- Antigen synonyms:MIS14|C1orf48|DC8
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: NSL1 (NSL1, MIS12 kinetochore complex component)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: