Anti-GET4 Rabbit Polyclonal Antibody
AVIVARP61197_P050
- Antibody type:Primary
- Antigen name:Golgi to ER traffic protein 4
- Antigen symbol:GET4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:51608
- Antigen synonyms:TRC35|CEE|CGI-20|C7orf20
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: GET4 (golgi to ER traffic protein 4 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human