Anti-AGK Rabbit Polyclonal Antibody
AVIVARP57169_P050
- Antibody type:Primary
- Antigen name:acylglycerol kinase
- Antigen symbol:AGK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:55750
- Antigen synonyms:MTDPS10|CATC5|MULK|CTRCT38
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: AGK (acylglycerol kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: