Anti-POPDC3 Rabbit Polyclonal Antibody
AVIVARP49709_P050
- Antibody type:Primary
- Antigen name:Popeye domain containing 3
- Antigen symbol:POPDC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:78977
- Antigen synonyms:AA682104|Pop3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: POPDC3 (popeye domain containing 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: