Order Entry
Spain
ContactUsLinkComponent
 
Anti-POC5 Rabbit Polyclonal Antibody
  AVIVARP49057_P050
undefined
Anti-POC5 Rabbit Polyclonal Antibody
  AVIVARP49057_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    POC5 centriolar protein
  • Antigen symbol:
    POC5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Pig,
    Human,
    Bovine,
    Guinea pig,
    Dog,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Gene ID:
    134359
  • Antigen synonyms:
    C5orf37
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: POC5 (POC5 centriolar protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Pig