Order Entry
Spain
ContactUsLinkComponent
 
Anti-PWWP2B Rabbit Polyclonal Antibody
  AVIVARP49005_P050
undefined
Anti-PWWP2B Rabbit Polyclonal Antibody
  AVIVARP49005_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    PWWP domain containing 2B
  • Antigen symbol:
    PWWP2B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    170394
  • Antigen synonyms:
    bA432J24.1|PWWP2|pp8607
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: PWWP2B (PWWP domain containing 2B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: