Anti-ACP6 Rabbit Polyclonal Antibody
Catalog # AVIVARP48363_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Acid Phosphatase 6
- Antigen symbol:ACP6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:51205
- Antigen synonyms:LPAP|PACPL1|ACPL1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ACP6 (acid phosphatase 6, lysophosphatidic)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: