Anti-RDH11 Rabbit Polyclonal Antibody
AVIVARP48350_P050
- Antibody type:Primary
- Antigen name:Retinol Dehydrogenase 11
- Antigen symbol:RDH11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:51109
- Antigen synonyms:ARSDR1, CGI82, HCBP12, MDT1, PSDR1, RALR1, RDJCSS, SCALD, SDR7C1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: RDH11 (retinol dehydrogenase 11 (all-trans/9-cis/11-cis))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: