Anti-CAP1 Rabbit Polyclonal Antibody
Catalog # AVIVARP48295_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:CAP, adenylate cyclase-associated protein 1 (yeast)
- Antigen symbol:CAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:10487
- Antigen synonyms:CAP|CAP1-PEN
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CAP1 (CAP, adenylate cyclase-associated protein 1 (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: