Anti-GPT Rabbit Polyclonal Antibody
Catalog # AVIVARP45711_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Glutamic-pyruvate Transaminase
- Antigen symbol:GPT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:2875
- Antigen synonyms:AAT1|GPT1|ALT1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: GPT (glutamic-pyruvate transaminase (alanine aminotransferase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: