Order Entry
Spain
ContactUsLinkComponent
 
Anti-MGST2 Rabbit Polyclonal Antibody
  AVIVARP45307_T100
undefined
Anti-MGST2 Rabbit Polyclonal Antibody
  AVIVARP45307_T100

 

  • Antibody type:
    Primary
  • Antigen name:
    microsomal glutathione S-transferase 2
  • Antigen symbol:
    MGST2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    4258
  • Antigen synonyms:
    MGST-II|GST2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: MGST2 (microsomal glutathione S-transferase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: