Anti-POMT1 Rabbit Polyclonal Antibody
AVIVARP44994_P050
- Antibody type:Primary
- Antigen name:protein-O-mannosyltransferase 1
- Antigen symbol:POMT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:10585
- Antigen synonyms:MDDGC1|RT|LGMD2K|MDDGA1|MDDGB1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: POMT1 (protein-O-mannosyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: