Anti-TMUB2 Rabbit Polyclonal Antibody
AVIVARP44980_P050
- Antibody type:Primary
- Antigen name:transmembrane and ubiquitin-like domain containing 2
- Antigen symbol:TMUB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:79089
- Antigen synonyms:FP2653
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: TMUB2 (transmembrane and ubiquitin-like domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: