Anti-ARMCX6 Rabbit Polyclonal Antibody
AVIVARP44584_T100
- Antibody type:Primary
- Antigen name:armadillo repeat containing, X-linked 6
- Antigen symbol:ARMCX6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:54470
- Antigen synonyms:GASP10
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: ARMCX6 (armadillo repeat containing, X-linked 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: