Anti-ZFYVE27 Rabbit Polyclonal Antibody
Catálogo # AVIVARP44427_T100
Proveedor: Aviva Systems Biology
Especificaciones
- Antibody type:Primary
- Antigen name:zinc finger, FYVE domain containing 27
- Antigen symbol:ZFYVE27
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:118813
- Antigen synonyms:SPG33|PROTRUDIN
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Especificaciones
Acerca de este artículo
Type: Primary
Antigen: ZFYVE27 (zinc finger, FYVE domain containing 27)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: