Anti-HNRNPF Rabbit Polyclonal Antibody
Catalog # AVIVARP40565_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:heterogeneous nuclear ribonucleoprotein F
- Antigen symbol:HNRNPF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Bovine,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:3185
- Antigen synonyms:HNRPF|OK/SW-cl.23|mcs94-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: HNRNPF (heterogeneous nuclear ribonucleoprotein F)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Rat