Anti-SNRPN Rabbit Polyclonal Antibody
Catalog # AVIVARP40450_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:small nuclear ribonucleoprotein polypeptide N
- Antigen symbol:SNRPN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:6638
- Antigen synonyms:RT-LI|SNRNP-N|SMN|PWCR|SM-D|sm-N|HCERN3|SNURF-SNRPN
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SNRPN (small nuclear ribonucleoprotein polypeptide N)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: