Anti-CIAO1 Rabbit Polyclonal Antibody
AVIVARP38516_P050
- Antibody type:Primary
- Antigen name:cytosolic iron-sulfur protein assembly 1
- Antigen symbol:CIAO1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:26371
- Antigen synonyms:AW210570|Wdr39
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: CIAO1 (cytosolic iron-sulfur assembly component 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: